Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101707.1 | NP_001095177.1 | 1 - 2 | 99 | 90 | |
GenBank mRNA | Y00763.1 | - | 99 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |