Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101713.1 | NP_001095183.1 | 6 - 7 | 653 | 96 | |
XM_008258923.1 | XP_008257145.1 | 6 - 7 | 548 | 96 | ||
GenBank mRNA | X54041.1 | - | 653 | 96 | ||
X54044.1 | - | 518 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons |