Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACRV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082084.1 | NP_001075553.1 | 2 | 352 | 78 | |
GenBank mRNA | AF251558.1 | - | 2 | 352 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |