Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082258.2 | NP_001075727.1 | 5 | 753 | 98 | |
GenBank mRNA | L42979.1 | - | 5 | 753 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |