Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082366.1 | NP_001075835.1 | 1 | 849 | 94 | |
GenBank mRNA | U40631.1 | - | 1 | 849 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |