Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082670.1 | NP_001076139.1 | 5 | 1048 | 92 | |
GenBank mRNA | M98764.1 | - | 5 | 1048 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |