Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082701.1 | NP_001076170.1 | 1 | 1350 | 83 | |
GenBank mRNA | U46069.1 | - | 1 | 1350 | 83 |
Gene Symbol: LOC100354065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719752.4 | XP_002719798.1 | 1 | 2073 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Oryctolagus cuniculus Both primers and probe map within a single exon |