Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164863.1 | NP_001158335.1 | 3 - 4 | 470 | 83 | |
XM_008252716.2 | XP_008250938.1 | 4 - 5 | 1019 | 83 | ||
GenBank mRNA | EU731009.1 | - | 3 - 4 | 470 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |