Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082195.2 | NP_001075664.1 | 2 | 1069 | 70 | |
GenBank mRNA | D16545.1 | - | 2 | 1070 | 70 | |
M85233.1 | - | 1 | 1053 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |