Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082638.1 | NP_001076107.1 | 1 - 2 | 67 | 99 | |
GenBank mRNA | AF451328.1 | - | 1 - 2 | 67 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |