Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163067.1 | NP_001156539.1 | 3 - 4 | 497 | 94 | |
XM_008274050.2 | XP_008272272.1 | 3 - 4 | 486 | 94 | ||
XM_008274053.2 | XP_008272275.1 | 3 - 4 | 275 | 94 | ||
XM_008274060.2 | XP_008272282.1 | 3 - 4 | 388 | 94 | ||
XM_017350167.1 | XP_017205656.1 | 4 - 5 | 487 | 94 | ||
GenBank mRNA | AY165034.1 | - | 3 - 4 | 497 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |