Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259599.2 | XP_008257821.1 | 16 - 17 | 2953 | 104 | |
XM_017343367.1 | XP_017198856.1 | 16 - 17 | 2317 | 104 | ||
GenBank mRNA | AF184165.1 | - | 5 - 6 | 419 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |