Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719452.3 | XP_002719498.1 | 4 | 1085 | 100 | |
XM_008271640.2 | XP_008269862.1 | 4 | 1203 | 100 | ||
GenBank mRNA | DQ786778.1 | - | 1 | 249 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |