Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099959.2 | NP_001093429.1 | 694 | 80 | ||
XM_008251492.2 | XP_008249714.1 | 1569 | 80 | |||
GenBank mRNA | AM236594.1 | - | 629 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Oryctolagus cuniculus |