Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101715.2 | NP_001095185.1 | 5 - 6 | 656 | 65 | |
GenBank mRNA | X64300.1 | - | 656 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |