Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163116.1 | NP_001156588.1 | 1014 | 64 | ||
GenBank mRNA | EU095941.1 | - | 1014 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus |