Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170871.1 | NP_001164342.1 | 3 - 4 | 406 | 87 | |
GenBank mRNA | AJ002388.1 | - | 3 - 4 | 406 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |