Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171113.1 | NP_001164584.1 | 4 - 5 | 696 | 116 | |
GenBank mRNA | K01521.1 | - | 4 - 5 | 607 | 116 | |
M19137.1 | - | 4 - 5 | 696 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |