Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171255.1 | NP_001164726.1 | 1 - 2 | 116 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |