Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171268.2 | NP_001164739.1 | 1071 | 90 | ||
GenBank mRNA | J05034.1 | - | 1072 | 90 | ||
M19139.1 | - | 1094 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus |