Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP3A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171268.2 | NP_001164739.1 | 265 | 103 | ||
GenBank mRNA | J05034.1 | - | 266 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus |