Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171285.1 | NP_001164756.1 | 3 - 4 | 350 | 85 | |
XM_008261385.2 | XP_008259607.1 | 5 - 6 | 775 | 85 | ||
XM_008261386.2 | XP_008259608.1 | 5 - 6 | 808 | 85 | ||
XM_017343649.1 | XP_017199138.1 | 5 - 6 | 476 | 85 | ||
XM_017343650.1 | XP_017199139.1 | 5 - 6 | 771 | 85 | ||
XM_017343651.1 | XP_017199140.1 | 5 - 6 | 804 | 85 | ||
XM_017343652.1 | XP_017199141.1 | 5 - 6 | 481 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |