Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171285.1 | NP_001164756.1 | 4 - 5 | 476 | 95 | |
XM_008261385.2 | XP_008259607.1 | 6 - 7 | 901 | 95 | ||
XM_008261386.2 | XP_008259608.1 | 6 - 7 | 934 | 95 | ||
XM_017343649.1 | XP_017199138.1 | 6 - 7 | 602 | 95 | ||
XM_017343650.1 | XP_017199139.1 | 6 - 7 | 897 | 95 | ||
XM_017343651.1 | XP_017199140.1 | 6 - 7 | 930 | 95 | ||
XM_017343652.1 | XP_017199141.1 | 6 - 7 | 607 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |