Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171285.1 | NP_001164756.1 | 5 - 6 | 613 | 93 | |
XM_008261385.2 | XP_008259607.1 | 7 - 8 | 1038 | 93 | ||
XM_008261386.2 | XP_008259608.1 | 7 - 8 | 1071 | 93 | ||
XM_017343649.1 | XP_017199138.1 | 7 - 8 | 739 | 93 | ||
XM_017343650.1 | XP_017199139.1 | 7 - 8 | 1034 | 93 | ||
XM_017343651.1 | XP_017199140.1 | 7 - 8 | 1067 | 93 | ||
XM_017343652.1 | XP_017199141.1 | 7 - 8 | 744 | 93 | ||
XM_017343653.1 | XP_017199142.1 | 2 - 3 | 158 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |