Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171309.1 | NP_001164780.1 | 1 - 2 | 315 | 80 | |
XM_008268814.2 | XP_008267036.1 | 2 - 3 | 877 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |