Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171325.1 | NP_001164796.1 | 4 - 5 | 1101 | 83 | |
XM_008273038.2 | XP_008271260.1 | 6 - 7 | 1387 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |