Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171332.1 | NP_001164803.1 | 4 - 5 | 540 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |