Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171345.1 | NP_001164816.1 | 1 - 2 | 91 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |