Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171413.2 | NP_001164884.1 | 1208 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus |