Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171483.1 | NP_001164954.1 | 1 - 2 | 312 | 52 | |
XM_017343152.1 | XP_017198641.1 | 1 - 2 | 406 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |