Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199820.1 | NP_001186749.1 | 221 | 63 | ||
GenBank mRNA | EB379496.1 | - | 200 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus |