Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145872.2 | XP_015001358.1 | 1080 | 82 | ||
XM_028852865.1 | XP_028708698.1 | 801 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |