Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3HDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193954.3 | NP_001180883.1 | 606 | 83 | ||
XM_015122274.2 | XP_014977760.1 | 641 | 83 | |||
XM_028838408.1 | XP_028694241.1 | 695 | 83 | |||
XM_028838409.1 | XP_028694242.1 | 567 | 83 | |||
XM_028838410.1 | XP_028694243.1 | 695 | 83 | |||
GenBank mRNA | JU322645.1 | - | 613 | 83 | ||
JU473157.1 | - | 613 | 83 | |||
JV043252.1 | - | 615 | 83 | |||
JV528921.1 | - | 630 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |