Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265680.1 | NP_001252609.1 | 1453 | 78 | ||
XM_015130559.2 | XP_014986045.2 | 3083 | 78 | |||
XM_015130560.2 | XP_014986046.2 | 3083 | 78 | |||
XM_028843161.1 | XP_028698994.1 | 2059 | 78 | |||
GenBank mRNA | JV566976.1 | - | 1531 | 78 | ||
JV667261.1 | - | 1453 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |