Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114030.4 | XP_001114030.1 | 1739 | 77 | ||
XM_015119835.2 | XP_014975321.1 | 1744 | 77 | |||
GenBank mRNA | JV607500.1 | - | 79 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |