Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258195.1 | NP_001245124.1 | 920 | 61 | ||
XM_028837742.1 | XP_028693575.1 | 912 | 61 | |||
GenBank mRNA | JU328027.1 | - | 706 | 61 | ||
JU475473.1 | - | 920 | 61 | |||
JV636131.1 | - | 802 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |