Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266232.1 | NP_001253161.1 | 395 | 113 | ||
GenBank mRNA | JU473042.1 | - | 395 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |