Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRUNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151853.2 | XP_015007339.1 | 1352 | 68 | ||
XM_015151860.2 | XP_015007346.1 | 1352 | 68 | |||
XM_015151871.2 | XP_015007357.1 | 538 | 68 | |||
XM_028829239.1 | XP_028685072.1 | 311 | 68 | |||
GenBank mRNA | JU335728.1 | - | 653 | 68 | ||
JU476613.1 | - | 658 | 68 | |||
JV047804.1 | - | 658 | 68 | |||
JV720214.1 | - | 638 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |