Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RORA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100971.4 | XP_001100971.2 | 994 | 102 | ||
XM_015142369.2 | XP_014997855.2 | 1322 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |