Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151823.2 | XP_015007309.2 | 4988 | 67 | ||
XM_015151825.2 | XP_015007311.2 | 4718 | 67 | |||
XM_015151826.2 | XP_015007312.2 | 4947 | 67 | |||
XM_028829509.1 | XP_028685342.1 | 4103 | 67 | |||
XM_028829510.1 | XP_028685343.1 | 4745 | 67 | |||
GenBank mRNA | JU333663.1 | - | 4209 | 67 | ||
JV646772.1 | - | 4047 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |