Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261629.1 | NP_001248558.1 | 380 | 81 | ||
XM_028843747.1 | XP_028699580.1 | 393 | 81 | |||
GenBank mRNA | JU333826.1 | - | 380 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |