Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H4orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193510.2 | NP_001180439.1 | 197 | 145 | ||
XM_015138946.2 | XP_014994432.1 | 669 | 145 | |||
XM_015138947.2 | XP_014994433.1 | 589 | 145 | |||
XM_028847953.1 | XP_028703786.1 | 805 | 145 | |||
XM_028847954.1 | XP_028703787.1 | 617 | 145 | |||
GenBank mRNA | JV707081.1 | - | 351 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |