Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU334587.1 | - | 6443 | 63 | ||
JV715523.1 | - | 6466 | 63 | |||
JV715524.1 | - | 6472 | 63 |
Gene Symbol: CLVS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265738.1 | NP_001252667.1 | 1282 | 63 | ||
GenBank mRNA | JV564997.1 | - | 1282 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |