Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260968.1 | NP_001247897.1 | 396 | 154 | ||
XM_015121722.2 | XP_014977208.1 | 578 | 154 | |||
XM_028837767.1 | XP_028693600.1 | 578 | 154 | |||
GenBank mRNA | JU319796.1 | - | 397 | 154 | ||
JU473617.1 | - | 397 | 154 | |||
JV043311.1 | - | 396 | 154 | |||
JV638127.1 | - | 402 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |