Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194497.1 | NP_001181426.1 | 348 | 124 | ||
XM_002804589.3 | XP_002804635.1 | 1045 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |