Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261678.1 | NP_001248607.1 | 1206 | 80 | ||
XM_015115807.2 | XP_014971293.1 | 1429 | 80 | |||
GenBank mRNA | JU474267.1 | - | 1206 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |