Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109811.3 | NP_001103281.1 | 481 | 74 | ||
XM_015116594.2 | XP_014972080.1 | 521 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |