Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260704.1 | NP_001247633.1 | 476 | 99 | ||
GenBank mRNA | JU336763.1 | - | 476 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |