Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261052.1 | NP_001247981.1 | 583 | 94 | ||
GenBank mRNA | JV048562.1 | - | 583 | 94 | ||
JV635822.1 | - | 697 | 94 |
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU348842.1 | - | 9421 | 94 | ||
JU493948.1 | - | 9377 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |