Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194509.2 | NP_001181438.1 | 306 | 85 | ||
XM_001111442.4 | XP_001111442.1 | 597 | 85 | |||
XM_028845066.1 | XP_028700899.1 | 333 | 85 | |||
XM_028845067.1 | XP_028700900.1 | 333 | 85 | |||
GenBank mRNA | JU320778.1 | - | 246 | 85 | ||
JU473032.1 | - | 251 | 85 | |||
JV043521.1 | - | 250 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |